Learn More
Chem Service Inc Chlorothalonil,1897-45-6,250MG
SDP
Catalog No. NC9865647
Encompass_Preferred
Molecular Formula C8Cl4N2; Purity 99.5; Molecular Weight 265.89; Boiling Point 350 °C; Melting Point 250 - 251 °C; Synonyms: Tetrachloroisophthalonitrile
Catalog No. NC9865647 Supplier Chem Service Inc Supplier No. N11454250MG
Molecular Formula C8Cl4N2; Purity 99.5; Molecular Weight 265.89; Boiling Point 350 °C; Melting Point 250 - 251 °C; Synonyms: Tetrachloroisophthalonitrile