Learn More
Medchemexpress LLC Chlorothalonil (Standard) | 1897-45-6 | 100 MG
SDP
Catalog No. 5000420176
Encompass_Preferred
Chlorothalonil (Standard) is an analytical standard of Chlorothalonil, intended for research and analytical applications. It functions as a broad-spectrum foliar fungicide with oral activity, used to combat fungal diseases in vegetable and crop leaves.
  • Used as an analytical standard for Chlorothalonil
  • Intended for research and analytical applications
  • Functions as a broad-spectrum foliar fungicide
  • Exhibits oral activity
  • Combats fungal diseases in vegetable and crop leaves
  • Can alter the microbial community in the soil
  • Inhibits spermatogenesis
  • Can cause intestinal epithelial barrier dysfunction and fetal toxicity
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-20176 Supplier Medchemexpress LLC Supplier No. HYN6625R100MG
Chlorothalonil (Standard) is an analytical standard of Chlorothalonil, intended for research and analytical applications. It functions as a broad-spectrum foliar fungicide with oral activity, used to combat fungal diseases in vegetable and crop leaves.
  • Used as an analytical standard for Chlorothalonil
  • Intended for research and analytical applications
  • Functions as a broad-spectrum foliar fungicide
  • Exhibits oral activity
  • Combats fungal diseases in vegetable and crop leaves
  • Can alter the microbial community in the soil
  • Inhibits spermatogenesis
  • Can cause intestinal epithelial barrier dysfunction and fetal toxicity
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS