Learn More
Apexbio Technology LLC Chlorothiazide, 100mg Cas# 58-94-6
SDP
Supplier:  Apexbio Technology LLC A8397100
Encompass_Preferred
Chlorothiazide is an inhibitor of carbonic anhydrase and is slightly less potent than acetazolamide. 58-94-6. MFCD00058576 Chlorothiazide is an inhibitor of carbonic anhydrase and is slightly less potent than acetazolamide. This compound has been shown to block reabsorption of sodium and chloride ions. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8595