Learn More
Apexbio Technology LLC Chlorothiazide 58-94-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062382
Encompass_Preferred
Chlorothiazide is a sulfonamide-derived diuretic compound known for its inhibitory action on carbonic anhydrase enzymes It functions by reducing sodium and chloride ion reabsorption via blockade of renal tubular transport mechanisms which subsequently promotes increased urine output Structurally Chlorothiazide contains a benzothiadiazine ring system contributing to its specific interaction with renal transport channels and carbonic anhydrase enzymes In biomedical research Chlorothiazide serves as a pharmacological tool to investigate ion transport pathways renal physiologic processes and mechanisms underlying fluid balance regulation Additionally it is utilized in experimental models studying electrolyte homeostasis and disorders associated with abnormal sodium or chloride retention
Catalog No. 50-000-62382 Supplier Apexbio Technology LLC Supplier No. A83975.1
Chlorothiazide is a sulfonamide-derived diuretic compound known for its inhibitory action on carbonic anhydrase enzymes It functions by reducing sodium and chloride ion reabsorption via blockade of renal tubular transport mechanisms which subsequently promotes increased urine output Structurally Chlorothiazide contains a benzothiadiazine ring system contributing to its specific interaction with renal transport channels and carbonic anhydrase enzymes In biomedical research Chlorothiazide serves as a pharmacological tool to investigate ion transport pathways renal physiologic processes and mechanisms underlying fluid balance regulation Additionally it is utilized in experimental models studying electrolyte homeostasis and disorders associated with abnormal sodium or chloride retention