Learn More
Medchemexpress LLC Chlorothiazide | 58-94-6 | 99.0% | 25 MG
SDP
Catalog No. 5000451005
Encompass_Preferred
Chlorothiazide is an orally active diuretic and anti-hypertensive agent intended for research use only.
  • Orally active diuretic and anti-hypertensive agent
  • For research use only
  • White to off-white solid appearance
  • Plasma half-life of 45-120 min
  • Biological half-life of nearly 10 hours
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-51005 Supplier Medchemexpress LLC Supplier No. 25MGHYB0224R
Add to Cart
Add to Cart
Chlorothiazide is an orally active diuretic and anti-hypertensive agent intended for research use only.
  • Orally active diuretic and anti-hypertensive agent
  • For research use only
  • White to off-white solid appearance
  • Plasma half-life of 45-120 min
  • Biological half-life of nearly 10 hours
  • Soluble in DMSO at 100 mg/mL