Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorothiazide | 58-94-6 | MFCD00058576 | 99.3% | 295.72 g/mol | C7H6ClN3O4S2 | 10 MG
SDP

Catalog No. 5000298264
Encompass_Preferred

Chlorothiazide (standard) is an analytical reference standard of the thiazide diuretic chlorothiazide supplied as a high-purity material for analytical and research use. It is intended for method development, calibration, and qualitative or quantitative analysis by HPLC, GC, and MS.

  • High purity: 99.3%.
  • Intended for HPLC, GC, and MS applications.
  • Supplied in small, weighed quantities suitable for reference use.
  • CAS number 58-94-6.
  • Molecular weight 295.72 g/mol.

Catalog No. 50-002-98264 Supplier Medchemexpress LLC Supplier No. HYB0224R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorothiazide (standard) is an analytical reference standard of the thiazide diuretic chlorothiazide supplied as a high-purity material for analytical and research use. It is intended for method development, calibration, and qualitative or quantitative analysis by HPLC, GC, and MS.

  • High purity: 99.3%.
  • Intended for HPLC, GC, and MS applications.
  • Supplied in small, weighed quantities suitable for reference use.
  • CAS number 58-94-6.
  • Molecular weight 295.72 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.