Learn More
Medchemexpress LLC Chlorothiazide (Standard) | 58-94-6 | 99.3% | 100 MG
SDP
Catalog No. 5000405653
Encompass_Preferred
Chlorothiazide (Standard) is the analytical standard of Chlorothiazide. This product is intended for research and analytical applications. Chlorothiazide is an orally active diuretic and anti-hypertensive agent. The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Analytical standard
  • Intended for research and analytical applications
  • Orally active diuretic
  • Anti-hypertensive agent
  • Reference standard supplied assay
  • Commonly used in HPLC, GC and MS
Catalog No. 50-004-05653 Supplier Medchemexpress LLC Supplier No. HYB0224R100MG
Chlorothiazide (Standard) is the analytical standard of Chlorothiazide. This product is intended for research and analytical applications. Chlorothiazide is an orally active diuretic and anti-hypertensive agent. The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Analytical standard
  • Intended for research and analytical applications
  • Orally active diuretic
  • Anti-hypertensive agent
  • Reference standard supplied assay
  • Commonly used in HPLC, GC and MS