Learn More
Medchemexpress LLC Chlorothiazide (Standard) | 58-94-6 | 99.3% | 50 MG
SDP
Catalog No. 5000406131
Encompass_Preferred
Chlorothiazide (Standard) is the analytical standard of Chlorothiazide, intended for research and analytical applications. It functions as an orally active diuretic and anti-hypertensive agent.
  • Analytical standard
  • Intended for research and analytical applications
  • Orally active diuretic and anti-hypertensive agent
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS experiments
  • For research use only
Catalog No. 50-004-06131 Supplier Medchemexpress LLC Supplier No. HYB0224R50MG
Chlorothiazide (Standard) is the analytical standard of Chlorothiazide, intended for research and analytical applications. It functions as an orally active diuretic and anti-hypertensive agent.
  • Analytical standard
  • Intended for research and analytical applications
  • Orally active diuretic and anti-hypertensive agent
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS experiments
  • For research use only