Learn More
Chlorotitanium triisopropoxide, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS | 20717-86-6, 75-09-2 |
| Molecular Formula | C9H21ClO3Ti |
| Linear Formula | {(CH3)2CHO}3TiCl |
| Quantity | 100mL |
| MDL Number | MFCD00009861 |
| Fieser | 13,213; 14,87 |
| Merck Index | 15, 2174 |
| Synonym | Chlorotriisopropyloxytitane |
| Formula Weight | 260.61 |
| Chemical Name or Material | Chlorotitanium triisopropoxide |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. Suspected of causing cancer. Contact with water liberates toxic gas. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.