Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorotoluron | 15545-48-9 | 99.9% | 250 MG
SDP

Catalog No. 5000414236
Encompass_Preferred

Chlorotoluron (Chlortoluron) is a substituted phenylurea herbicide, widely used for selective weed control in cereal crops and is an environmental pollutant.

  • Molecular weight: 212.68
  • Chemical formula: C10H13ClN2O
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Soluble in DMSO (50 mg/mL)
  • Used as a herbicide
  • Also classified as a metabolic enzyme/protease

Catalog No. 50-004-14236 Supplier Medchemexpress LLC Supplier No. 250MGHYB2023
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorotoluron (Chlortoluron) is a substituted phenylurea herbicide, widely used for selective weed control in cereal crops and is an environmental pollutant.

  • Molecular weight: 212.68
  • Chemical formula: C10H13ClN2O
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Soluble in DMSO (50 mg/mL)
  • Used as a herbicide
  • Also classified as a metabolic enzyme/protease

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.