Learn More
CPC Scientific H-Met-Cys-Met-Pro-Cys-Phe-Thr-Thr-Asp-His-Gln-Met-Ala-Arg-Lys-Cys-Asp-Asp-Cys-Cys-Gly-Gly-Lys-Gly-Arg-Gly-Lys-Cys-Tyr-Gly-Pro-Gln-Cys-Leu-Cys-Arg-NH2 (trifluoroacetate salt)
(Disulfide Bridge: Cys1-Cys4,Cys2-Cys6,Cys3-Cys7,Cys5-Cys8) 0.5MG
SDP
Supplier:  CPC Scientific CLTX001B
Encompass
Chlorotoxin is a chloride channel blocker originally isolate from the venom of the deathstalker scorpion. This 36-mer peptide also selectively binds to glioma cells which has made it a candiate for cancer therapeutics.SEQUENCE: H-Met-Cys-Met-Pro-Cys-Phe-Thr-Thr-Asp-His-Gln-Met-Ala-Arg-Lys-Cys-Asp-Asp-Cys-Cys-Gly-Gly-Lys-Gly-Arg-Gly-Lys-Cys-Tyr-Gly-Pro-Gln-Cys-Leu-Cys-Arg-NH2 (trifluoroacetate salt)(Disulfide Bridge: Cys1-Cys4,Cys2-Cys6,Cys3-Cys7,Cys5-Cys8)ONE-LETTER SEQUENCE: MCMPCFTTDHQMARKCDDCCGGKGRGKCYGPQCLCR-NH2MOLECULAR FORMULA: C158H249N53O47S11MOLECULAR WEIGHT: 3995.77STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [163515-35-3]SYNONYMS: RESEARCH AREA: ToxinREFERENCES:DeBin JA, Strichartz GR. Toxicon 29 (11): 1403, 8, (1991).Deshane J, Garner CC, Sontheimer H . J. Biol. Chem. 278 (6): 4135, 44, (2003).SKU(s): CLTX-001A, CLTX-001B
Catalog No. 50-210-9128