Learn More
Enzo Life Sciences Chlorotoxin (500µg). CAS: 163515-35-3
SDP
Catalog No. 502009613
Encompass_Preferred
Potent neurotoxin. Small conductance Cl- channel blocker. Highly specific marker for glioma cells and tumors of neuroectodermal origin. Inhibits the activity of matrix metalloproteinase-2 (MMP-2). Purity: ≥98% (TLC, HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9613 Supplier Enzo Life Sciences Supplier No. ALX630069C500
Add to Cart
Add to Cart
Potent neurotoxin. Small conductance Cl- channel blocker. Highly specific marker for glioma cells and tumors of neuroectodermal origin. Inhibits the activity of matrix metalloproteinase-2 (MMP-2). Purity: ≥98% (TLC, HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.