Learn More
Medchemexpress LLC HY-P0173B 1mg Medchemexpress, Chlorotoxin (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0173B1MG
Encompass_Preferred
Medchemexpress, HY-P0173B 1mg Chlorotoxin (TFA) CAS: Chlorotoxin TFA is a peptide isolated from the venom of the scorpion Leiurus quinquestriatus, acts as a chloride channel blocker [1] . Anti-cancer activity [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4039