Learn More
Medchemexpress LLC HY-B2158 5mg Medchemexpress, Chlorotrianisene CAS:569-57-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB21585MG
Encompass_Preferred
Medchemexpress, HY-B2158 5mg Chlorotrianisene CAS:569-57-3 Chlorotrianisene is a long-acting non-steroidal estrogen and an orally active estrogen receptor modulator. Chlorotrianisene exhibits antiestrogenic activity. Chlorotrianisene potently inhibits the enzyme COX-1 and inhibits platelet aggregation in whole blood. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2727