Learn More
Strem, An Ascensus Company CAS# 15529-90-5. 1g. Chlorotriethylphosphinegold(I), 99%. MFCD00013205
SDP
Supplier:  Strem, An Ascensus Company 7907501G
Encompass
CAS# 15529-90-5. 1g. Chlorotriethylphosphinegold(I), 99%. MFCD00013205. Molecular Weight: 350.57. Molecular Formula: AuClP(C2H5)3. Color/form: white to pink xtl. Strem# 79-0750. http://www.strem.com/catalog/v/79-0750/
Catalog No. 50-901-13395