Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorotriethylsilane | 994-30-9 | 98.3% | 150.72 g/mol | C6H15ClSi | 100 G
SDP

Catalog No. 5000208510
Encompass_Preferred

Chlorotriethylsilane (triethylsilyl chloride) is an organosilicon reagent used in organic synthesis to introduce the triethylsilyl (TES) protecting group and to perform silylation reactions. It is supplied as a colorless to light yellow liquid with typical purity of 98.3% and is available in multiple packaging sizes.

  • Used for silylation and introducing triethylsilyl (TES) protecting groups.
  • Colorless to light yellow liquid.
  • Typical purity 98.3%.
  • Molecular formula C6H15ClSi, molecular weight 150.72 g/mol.
  • Available in 25 g, 50 g, 100 g, and 500 g packaging.
  • Certificate of analysis available.

Catalog No. 50-002-08510 Supplier Medchemexpress LLC Supplier No. HYW040306100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorotriethylsilane (triethylsilyl chloride) is an organosilicon reagent used in organic synthesis to introduce the triethylsilyl (TES) protecting group and to perform silylation reactions. It is supplied as a colorless to light yellow liquid with typical purity of 98.3% and is available in multiple packaging sizes.

  • Used for silylation and introducing triethylsilyl (TES) protecting groups.
  • Colorless to light yellow liquid.
  • Typical purity 98.3%.
  • Molecular formula C6H15ClSi, molecular weight 150.72 g/mol.
  • Available in 25 g, 50 g, 100 g, and 500 g packaging.
  • Certificate of analysis available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.