Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorotriethylsilane | 994-30-9 | MFCD00000507 | ≥97.0% | 150.72 g/mol | C6H15ClSi | 25 G
SDP

Catalog No. 5000209747
Encompass_Preferred

Chlorotriethylsilane (triethylsilyl chloride) is an organosilicon reagent used in organic synthesis, commonly employed as a silylating agent and protecting group. It is supplied as a colorless to light yellow liquid with documented purity and storage recommendations for research use.

  • Used as a silylating reagent and protecting group in organic synthesis.
  • Supplied as a colorless to light yellow liquid; available in 25 g and larger sizes.
  • Molecular formula C6H15ClSi; CAS number 994-30-9.
  • Purity ≥97.0% (NMR) as verified by certificate of analysis.
  • Intended for research use only; not for clinical, diagnostic, or food use.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-09747 Supplier Medchemexpress LLC Supplier No. 25GHYW040306
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorotriethylsilane (triethylsilyl chloride) is an organosilicon reagent used in organic synthesis, commonly employed as a silylating agent and protecting group. It is supplied as a colorless to light yellow liquid with documented purity and storage recommendations for research use.

  • Used as a silylating reagent and protecting group in organic synthesis.
  • Supplied as a colorless to light yellow liquid; available in 25 g and larger sizes.
  • Molecular formula C6H15ClSi; CAS number 994-30-9.
  • Purity ≥97.0% (NMR) as verified by certificate of analysis.
  • Intended for research use only; not for clinical, diagnostic, or food use.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.