Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorotriethylsilane | 994-30-9 | MFCD00000507 | 98.3% | 150.72 g/mol | C6H15ClSi | 500 G
SDP

Catalog No. 5000208509
Encompass_Preferred

Chlorotriethylsilane (chloro(triethyl)silane) is an organosilicon reagent used in organic synthesis for silylation and protecting-group chemistry. It is supplied as a research-grade liquid with supporting SDS and COA documentation available.

  • Used for silylation and protecting-group formation in organic synthesis.
  • Chemical formula C6H15ClSi; molecular weight 150.72 g/mol.
  • Purity 98.33%.
  • Available in a 500 G pack size.
  • SDS and COA available for safe handling and quality verification.

Catalog No. 50-002-08509 Supplier Medchemexpress LLC Supplier No. HYW040306500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorotriethylsilane (chloro(triethyl)silane) is an organosilicon reagent used in organic synthesis for silylation and protecting-group chemistry. It is supplied as a research-grade liquid with supporting SDS and COA documentation available.

  • Used for silylation and protecting-group formation in organic synthesis.
  • Chemical formula C6H15ClSi; molecular weight 150.72 g/mol.
  • Purity 98.33%.
  • Available in a 500 G pack size.
  • SDS and COA available for safe handling and quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.