Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorotriisobutylsilane

Catalog No. 2910135G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-101-35G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 29-101-35G Supplier Sigma Aldrich Supplier No. 2910135G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorotriisobutylsilane can be used as an intermediate in the synthesis of:Silicon (IV) phthalocyanines, which are used as photosensitizers for the photodynamicinactivationofbacteria.Poly(4-alkylthiazole) polymers exhibiting optical and electronic properties.Bis(trialkylsilyl oxide) based silicon phthalocyanines applicable as ternary additives in organic photovoltaics.

Specifications

CAS 204777-78-6
Molecular Formula C34H42N2O6
Refractive Index n20/D 1.448
Linear Formula C34H42N2O6
MDL Number MFCD01631658
Quantity 1 g
Synonym Triisobutylchlorosilane; Triisobutylsilyl chloride
Molecular Weight (g/mol) 574.71 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.