Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorotriisopropoxytitanium(IV) solution

Catalog No. 252670100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-267-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-267-0100ML Supplier Sigma Aldrich Supplier No. 252670100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorotriisopropoxytitanium(IV) solution may be used as a precursor for the synthesis of ultralow density ceramic materials. The product may be used to mediate a reductive amination reaction of 5α-cholestane-3,7-dione to yield aryl aminocholestanes.

Specifications

CAS 20717-86-6
Density 0.775 g/mL (at 25°C)
Molecular Formula C9H21ClO3Ti
Linear Formula [(CH3)2 CHO]3 TiCl
Quantity 100 mL
MDL Number MFCD00009861
Molecular Weight (g/mol) 260.58
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.