Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorotrimethylsilane

Catalog No. 3865291L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
1 L
2 L
4 x 100 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-652-91L 1 L
38-652-9100ML 100 mL
38-652-925ML 25 mL
38-652-9 4 x 100 mL
38-652-92L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 38-652-91L Supplier Sigma Aldrich Supplier No. 3865291L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorotrimethylsilane

Specifications

CAS 75-77-4
Melting Point -40°C
Density 0.856 g/mL at 25°C
Boiling Point 57°C
Molecular Formula C3H9ClSi
Refractive Index n20/D 1.387
Linear Formula (CH3)3SiCl
MDL Number MFCD00000502
Quantity 1 L
Synonym TMSCl; TMCS; Trimethylchlorosilane; Trimethylsilyl chloride
Molecular Weight (g/mol) 108.64 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.