Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorotrimethylsilane Solution

Catalog No. 384410100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-441-0100ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 38-441-0100ML Supplier Sigma Aldrich Supplier No. 384410100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorotrimethylsilane (TMSCl) is a silylating agent employed in the silylation of alcohols, amines, carboxylic acids, thiols, and other functional groups. Silyl group serves as a protecting group for these original functional groups.Chlorotrimethylsilane (TMSCl) can be used as a reagent: To synthesize allenylsilane via carbolithiation of conjugated enynes with aryllithium. In the reductive coupling of aldehydes and ketones to produce corresponding 1,2 diols in the presence of zirconocene dichloride and magnesium. To prepare 4,4'sa-bis(trimethylsilyl)bicyclohexyl-2,2'sa-diene from 1,3-cyclohexadiene by reductive disilylation reaction.

Specifications

CAS 641-74-7
Molecular Formula C20H16Cl2Zr
Linear Formula C20H16Cl2Zr
MDL Number MFCD01314544
Quantity 1 g
Synonym Trimethylchlorosilane
Molecular Weight (g/mol) 418.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.