Learn More
Strem, An Ascensus Company CAS# 14243-64-2. 1g. Chlorotriphenylphosphinegold(I), 98+% (99.9+%-Au). MFCD00009588
SDP
Catalog No. 5090113400
Encompass
CAS# 14243-64-2. 1g. Chlorotriphenylphosphinegold(I), 98+% (99.9+%-Au). MFCD00009588. Molecular Weight: 494.71. Molecular Formula: AuClP(C6H5)3. Color/form: white xtl. Strem# 79-1000. http://www.strem.com/catalog/v/79-1000/
Catalog No. 50-901-13400 Supplier Strem, An Ascensus Company Supplier No. 7910001G
CAS# 14243-64-2. 1g. Chlorotriphenylphosphinegold(I), 98+% (99.9+%-Au). MFCD00009588. Molecular Weight: 494.71. Molecular Formula: AuClP(C6H5)3. Color/form: white xtl. Strem# 79-1000. http://www.strem.com/catalog/v/79-1000/