Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorotris(trimethylsilyl)silane 95.0+%, TCI America™
SDP

Catalog No. C24115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24115G Supplier TCI America Supplier No. C24115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5565-32-2
Molecular Formula C21H13NO4S2
Molecular Weight (g/mol) 407.46
MDL Number MFCD01321222
InChI Key CVCSUQKXXAAHNH-QGOAFFKASA-N
Synonym Tris(trimethylsilyl)silyl Chloride
PubChem CID 4641097
IUPAC Name 3-{[(5E)-4-oxo-3-phenyl-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}phenyl furan-2-carboxylate
SMILES O=C(OC1=CC=CC(\C=C2\SC(=S)N(C2=O)C2=CC=CC=C2)=C1)C1=CC=CO1

Specifications

Melting Point 46°C
Color White
Boiling Point 93°C
Quantity 5 g
UN Number 2925
Formula Weight 283.11
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Chlorotris(trimethylsilyl)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.