Learn More
Strem, An Ascensus Company CAS# 14694-95-2. 1g. Chlorotris(triphenylphosphine)rhodium(I), 99% WILKINSON'S CATALYST. MFCD00010016
SDP
Catalog No. 5090112159
Encompass
CAS# 14694-95-2. 1g. Chlorotris(triphenylphosphine)rhodium(I), 99% WILKINSON'S CATALYST. MFCD00010016. Molecular Weight: 925.23. Molecular Formula: RhCl(P(C6H5)3)3. Color/form: maroon xtl. Strem# 45-0650. http://www.strem.com/catalog/v/45-0650/
Catalog No. 50-901-12159 Supplier Strem, An Ascensus Company Supplier No. 4506501G
CAS# 14694-95-2. 1g. Chlorotris(triphenylphosphine)rhodium(I), 99% WILKINSON'S CATALYST. MFCD00010016. Molecular Weight: 925.23. Molecular Formula: RhCl(P(C6H5)3)3. Color/form: maroon xtl. Strem# 45-0650. http://www.strem.com/catalog/v/45-0650/