Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorotris(triphenylphosphine)copper | 15709-76-9 | MFCD00010270 | 98.0% | 885.86 g/mol | C54H45ClCuP3 | 1 G
SDP

Catalog No. 5000284152
Encompass_Preferred

Chlorotris(triphenylphosphine)copper (CuCl(TPP)3) is a copper(I) coordination complex supplied as a high-purity solid for research use. It is used in biochemical and medicinal chemistry studies where metal-ligand interactions and DNA binding are of interest, and has reported antibacterial, antifungal, antitumor, and antioxidant activity.

  • High purity (98.0%).
  • Molecular weight 885.86 g/mol.
  • Chemical formula C54H45ClCuP3.
  • Supplied in research pack sizes (1 g, 5 g, 10 g, 25 g).
  • Store at room temperature; protect from light.
  • Downloadable data sheet, safety data sheet, and certificate of analysis available.

Catalog No. 50-002-84152 Supplier Medchemexpress LLC Supplier No. HYW5825041G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorotris(triphenylphosphine)copper (CuCl(TPP)3) is a copper(I) coordination complex supplied as a high-purity solid for research use. It is used in biochemical and medicinal chemistry studies where metal-ligand interactions and DNA binding are of interest, and has reported antibacterial, antifungal, antitumor, and antioxidant activity.

  • High purity (98.0%).
  • Molecular weight 885.86 g/mol.
  • Chemical formula C54H45ClCuP3.
  • Supplied in research pack sizes (1 g, 5 g, 10 g, 25 g).
  • Store at room temperature; protect from light.
  • Downloadable data sheet, safety data sheet, and certificate of analysis available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.