Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorotris(triphenylphosphine)copper | 15709-76-9 | MFCD00010270 | 98.0% | 885.86 g·mol⁻¹ | C54H45ClCuP3 | 10 G
SDP

Catalog No. 5000284113
Encompass_Preferred

Chlorotris(triphenylphosphine)copper is a copper(I) coordination complex composed of a chloride ligand coordinated to three triphenylphosphine ligands. It is commonly used as a source of Cu(I) in organometallic synthesis and catalytic applications, and is supplied in laboratory pack sizes for research use.

  • Stable copper(I) complex suitable for synthesis and catalysis.
  • Soluble in common organic solvents; verify solubility for specific reactions.
  • High chemical purity for reproducible experimental results.
  • Available in multiple laboratory pack sizes for scale flexibility.
  • Molecular formula C54H45ClCuP3 supports stoichiometric calculations.

Catalog No. 50-002-84113 Supplier Medchemexpress LLC Supplier No. HYW58250410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorotris(triphenylphosphine)copper is a copper(I) coordination complex composed of a chloride ligand coordinated to three triphenylphosphine ligands. It is commonly used as a source of Cu(I) in organometallic synthesis and catalytic applications, and is supplied in laboratory pack sizes for research use.

  • Stable copper(I) complex suitable for synthesis and catalysis.
  • Soluble in common organic solvents; verify solubility for specific reactions.
  • High chemical purity for reproducible experimental results.
  • Available in multiple laboratory pack sizes for scale flexibility.
  • Molecular formula C54H45ClCuP3 supports stoichiometric calculations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.