Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorotris(triphenylphosphine)copper | 15709-76-9 | MFCD00010270 | 98.0% | 885.86 g/mol | C54H45ClCuP3 | 25 G
SDP

Catalog No. 5000284298
Encompass_Preferred

Chlorotris(triphenylphosphine)copper is a copper(I) coordination complex used in biochemical and pharmacological research as a DNA-targeted metal complex. It binds DNA through non-covalent interactions such as groove binding and has reported antibacterial, antifungal, antitumor, and antioxidant activity in experimental studies.

  • Copper(I) coordination complex useful for metal-DNA interaction studies.
  • Interacts with DNA via non-covalent (groove binding) mechanisms.
  • Reported antibacterial, antifungal, antitumor, and antioxidant activity.
  • Available as a powder in multiple pack sizes, including 25 G.
  • Molecular formula C54H45ClCuP3; molecular weight 885.86 g/mol.
  • Store at room temperature protected from light; solutions may be frozen for long-term storage.

Catalog No. 50-002-84298 Supplier Medchemexpress LLC Supplier No. HYW58250425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorotris(triphenylphosphine)copper is a copper(I) coordination complex used in biochemical and pharmacological research as a DNA-targeted metal complex. It binds DNA through non-covalent interactions such as groove binding and has reported antibacterial, antifungal, antitumor, and antioxidant activity in experimental studies.

  • Copper(I) coordination complex useful for metal-DNA interaction studies.
  • Interacts with DNA via non-covalent (groove binding) mechanisms.
  • Reported antibacterial, antifungal, antitumor, and antioxidant activity.
  • Available as a powder in multiple pack sizes, including 25 G.
  • Molecular formula C54H45ClCuP3; molecular weight 885.86 g/mol.
  • Store at room temperature protected from light; solutions may be frozen for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.