Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorotris(triphenylphosphine)copper | 15709-76-9 | MFCD00010270 | 98.0% | 885.86 g/mol | C54H45ClCuP3 | 5 G
SDP

Catalog No. 5000284153
Encompass_Preferred

Chlorotris(triphenylphosphine)copper is a copper(I) coordination complex in which a chloride ligand is coordinated to three triphenylphosphine ligands. It is used as a reagent or catalyst in organometallic and organic synthesis and for laboratory research applications that require copper(I)-phosphine chemistry.

  • Copper(I) triphenylphosphine chloride complex for research use.
  • High purity: 98.0%.
  • Molecular weight 885.86 g/mol.
  • Chemical formula C54H45ClCuP3.
  • CAS number 15709-76-9.
  • Available in small lab-scale packaging suitable for synthesis and characterization.

Catalog No. 50-002-84153 Supplier Medchemexpress LLC Supplier No. HYW5825045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorotris(triphenylphosphine)copper is a copper(I) coordination complex in which a chloride ligand is coordinated to three triphenylphosphine ligands. It is used as a reagent or catalyst in organometallic and organic synthesis and for laboratory research applications that require copper(I)-phosphine chemistry.

  • Copper(I) triphenylphosphine chloride complex for research use.
  • High purity: 98.0%.
  • Molecular weight 885.86 g/mol.
  • Chemical formula C54H45ClCuP3.
  • CAS number 15709-76-9.
  • Available in small lab-scale packaging suitable for synthesis and characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.