Learn More
Medchemexpress LLC HY-B0295 100mg Medchemexpress, Chloroxine CAS:773-76-2 Purity:>98%
SDP
Catalog No. 502082453
Encompass_Preferred
Medchemexpress, HY-B0295 100mg Chloroxine CAS:773-76-2 Chloroxine is one of the important 8-hydroxyquinoline derivative. Chloroxine has effective antibacterial, antifungal, antiprotozoal and antiamoebic activities, especially used in treating the intestinal amebiasis. Chloroxine is also used in the treatment of dandruff and seborrheic dermatitis of the scalp. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2453 Supplier Medchemexpress LLC Supplier No. HYB0295100MG
Medchemexpress, HY-B0295 100mg Chloroxine CAS:773-76-2 Chloroxine is one of the important 8-hydroxyquinoline derivative. Chloroxine has effective antibacterial, antifungal, antiprotozoal and antiamoebic activities, especially used in treating the intestinal amebiasis. Chloroxine is also used in the treatment of dandruff and seborrheic dermatitis of the scalp. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.