Learn More
Apexbio Technology LLC Chloroxine 773-76-2 50mg
SDP
Catalog No. 502240694
Encompass_Preferred
Chloroxine (CAS 773-76-2) is a small-molecule agent that exerts antimicrobial antifungal and antiprotozoal activity by chelating metal ions essential for microbial proliferation thereby disrupting intracellular metal ion homeostasis and impairing critical metabolic pathways in pathogens Chloroxine demonstrates inhibitory activity with reported IC50 values typically ranging from submicromolar to low micromolar concentrations depending on the microorganism and assay conditions Based on these pharmacological properties chloroxine holds research potential in studies of intestinal amoebiasis pathogenesis experimental therapeutic evaluations and investigations of antifungal agents targeting fungi involved in dandruff and seborrheic dermatitis
This item is not returnable. View return policy
Catalog No. 50-224-0694 Supplier Apexbio Technology LLC Supplier No. B191050
Chloroxine (CAS 773-76-2) is a small-molecule agent that exerts antimicrobial antifungal and antiprotozoal activity by chelating metal ions essential for microbial proliferation thereby disrupting intracellular metal ion homeostasis and impairing critical metabolic pathways in pathogens Chloroxine demonstrates inhibitory activity with reported IC50 values typically ranging from submicromolar to low micromolar concentrations depending on the microorganism and assay conditions Based on these pharmacological properties chloroxine holds research potential in studies of intestinal amoebiasis pathogenesis experimental therapeutic evaluations and investigations of antifungal agents targeting fungi involved in dandruff and seborrheic dermatitis