Learn More
Medchemexpress LLC Chloroxylenol | 88-04-0 | 99.5% | 100 G
SDP
Catalog No. 5000412668
Encompass_Preferred
Chloroxylenol is a broad-spectrum antibacterial agent that can be used to control bacteria, algae, fungi, and viruses. It inhibits the viability, proliferation, migration, invasion, and sphere formation, and induces apoptosis in HCT116 and SW480 cells by inhibiting the Wnt/β-catenin signaling pathway. It also inhibits nuclear translocation of β-catenin in HCT116 and SW480 cells and inhibits the growth of CRC organoids. In vivo, Chloroxylenol inhibits tumor growth in a MC38 cell xenograft model.
  • Broad-spectrum antibacterial agent
  • Effective against bacteria, algae, fungi, and viruses
  • Inhibits Wnt/β-catenin signaling pathway
  • Induces apoptosis in certain cancer cells
  • Inhibits tumor growth in xenograft models
  • Potential for anti-cancer activity in colorectal cancer
Catalog No. 50-004-12668 Supplier Medchemexpress LLC Supplier No. HYB1414100G
Chloroxylenol is a broad-spectrum antibacterial agent that can be used to control bacteria, algae, fungi, and viruses. It inhibits the viability, proliferation, migration, invasion, and sphere formation, and induces apoptosis in HCT116 and SW480 cells by inhibiting the Wnt/β-catenin signaling pathway. It also inhibits nuclear translocation of β-catenin in HCT116 and SW480 cells and inhibits the growth of CRC organoids. In vivo, Chloroxylenol inhibits tumor growth in a MC38 cell xenograft model.
  • Broad-spectrum antibacterial agent
  • Effective against bacteria, algae, fungi, and viruses
  • Inhibits Wnt/β-catenin signaling pathway
  • Induces apoptosis in certain cancer cells
  • Inhibits tumor growth in xenograft models
  • Potential for anti-cancer activity in colorectal cancer