Learn More
Medchemexpress LLC Chloroxylenol-d6 | 1407521-66-7 | 2.5 MG
SDP
Catalog No. 5000412579
Encompass_Preferred
Chloroxylenol-d6 is the deuterium-labeled form of Chloroxylenol, a broad-spectrum antimicrobial chemical compound. It is utilized to manage bacteria, algae, fungi, and viruses. This compound is for research use only and serves as a tracer and internal standard in various analytical techniques.
  • It is a deuterium labeled Chloroxylenol.
  • It exhibits broad-spectrum antimicrobial properties.
  • It is effective in controlling bacteria, algae, fungi, and viruses.
  • It can be used as a tracer in research applications.
  • It serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • The incorporation of stable heavy isotopes into drug molecules allows for their use as tracers for quantitation during the drug development process.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12579 Supplier Medchemexpress LLC Supplier No. HYB1414S2.5MG
Chloroxylenol-d6 is the deuterium-labeled form of Chloroxylenol, a broad-spectrum antimicrobial chemical compound. It is utilized to manage bacteria, algae, fungi, and viruses. This compound is for research use only and serves as a tracer and internal standard in various analytical techniques.
  • It is a deuterium labeled Chloroxylenol.
  • It exhibits broad-spectrum antimicrobial properties.
  • It is effective in controlling bacteria, algae, fungi, and viruses.
  • It can be used as a tracer in research applications.
  • It serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • The incorporation of stable heavy isotopes into drug molecules allows for their use as tracers for quantitation during the drug development process.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.