Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroxylenol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015739 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5739 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5739 Supplier MilliporeSigma Supelco Supplier No. PHR1478500MG
Add to Cart
Add to Cart

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 88-04-0
Linear Formula ClC6H2(CH3)2 OH
MDL Number MFCD00002324
Melting Point 114°C to 116°C
Synonym 4-Chloro-3,5-dimethylphenol; 4-Chloro-3,5-xylenol; 4-Chloro-sym-m-xylenol; PCMX
Type Certified Reference Material
Quantity 500 mg
Formula Weight 156.61
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.