Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroxylenol, USP, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18601637 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-637 12 kg Amber Glass Bottle
18-601-631 5 g Amber Glass Bottle
18-601-632 25 g Amber Glass Bottle
18-601-633 100 g Amber Glass Bottle
18-601-634 500 g Amber Glass Bottle
18-601-636 25 kg Amber Glass Bottle
18-601-635 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-601-637 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CH13612KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CH136, 88-04-0, C8H9ClO

Spectrum™ Chemical Chloroxylenol, USP is a broad spectrum antimicrobial chemical compound used to control bacteria, algae, fungi and virus. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 88-04-0
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
InChI Key OSDLLIBGSJNGJE-UHFFFAOYSA-N
IUPAC Name 4-chloro-3,5-dimethylphenol
SMILES CC1=CC(O)=CC(C)=C1Cl

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 12 kg
Percent Purity 98.5%
Grade USP
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.