Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloro(4-Cyanophenyl)[™-1-[(S)-2-(Dicyclohexylphosphino)Ferrocenyl]Ethyldiphenylphosphine]Nickel(II)

Catalog No. 900942250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-094-2250MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-094-2250MG Supplier Sigma Aldrich Supplier No. 900942250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Amination of aryl chlorides with ammonium salts.Nickel-Catalyzed Amination of Aryl Chlorides with Ammonia or Ammonium SaltsValorization of the Primary Building Blocks Ammonia and Acetone Featuring Pd- and Ni-Catalyzed Monoarylations

Specifications

Melting Point >300°C
Density 0.970 g/mL
Molecular Formula C61H68Cl2N2Ru -+ CH3NO2
Linear Formula C61H68Cl2N2Ru -+ CH3NO2
Quantity 100 mg
Synonym Bis(1-(2,6-diethylphenyl)-3,5,5-trimethyl-3-phenylpyrrolidin-2-ylidene)dichloro(3-phenyl-1H-inden-1-ylidene)ruthenium(II)
Molecular Weight (g/mol) 1062.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.