Learn More
Chlorphenamine N-Oxide Maleate, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111023005 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
11-102-3005 100 mg
1 options
Catalog No. 11-102-3005 Supplier MilliporeSigma Supelco Supplier No. PHR2105100MG
Only null left
Add to Cart
Add to Cart
Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY
Specifications
Grade Certified Reference Material
Quantity 100 mg