Learn More
Apexbio Technology LLC Chlorpheniramine Maleate 113-92-8 100mg
SDP
Catalog No. 502240526
Encompass_Preferred
Chlorpheniramine Maleate (CAS 113-92-8) is a synthetic small molecule that acts as a competitive antagonist at the histamine H1 receptor It exhibits high affinity for this receptor with an inhibitory concentration (IC50) of 12 nM effectively blocking histamine-mediated signal transduction Due to its well-characterized pharmacological profile chlorpheniramine maleate is widely employed in biomedical research to investigate histaminergic pathways allergic responses and the modulation of H1 receptor activity in various physiological and pathological contexts
This item is not returnable. View return policy
Catalog No. 50-224-0526 Supplier Apexbio Technology LLC Supplier No. B1556100
Chlorpheniramine Maleate (CAS 113-92-8) is a synthetic small molecule that acts as a competitive antagonist at the histamine H1 receptor It exhibits high affinity for this receptor with an inhibitory concentration (IC50) of 12 nM effectively blocking histamine-mediated signal transduction Due to its well-characterized pharmacological profile chlorpheniramine maleate is widely employed in biomedical research to investigate histaminergic pathways allergic responses and the modulation of H1 receptor activity in various physiological and pathological contexts