Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorpheniramine maleate, 99%

Catalog No. AAB2523814
Click to view available options
Quantity:
5 g
25 g

CAS: 113-92-8 | C20H23ClN2O4 | 390.86 g/mol

An antihistaminic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-92-8
Molecular Formula C20H23ClN2O4
Molecular Weight (g/mol) 390.86
MDL Number MFCD00069225
InChI Key DBAKFASWICGISY-BTJKTKAUNA-N
Synonym chlorpheniramine maleate, antagonate, allerclor, allergin, chlormene, alunex, carbinoxamide maleate, allergisan, -chlorpheniramine maleate salt, dl-2-p-chloro-a-2-dimethylamino ethylbenzyl pyridine bimaleate
PubChem CID 5281068
ChEBI CHEBI:3645
IUPAC Name [3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylazanium (2Z)-3-carboxyprop-2-enoate
SMILES OC(=O)\C=C/C([O-])=O.C[NH+](C)CCC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1

Specifications

Melting Point 130°C to 135°C
Quantity 25 g
Formula Weight 390.86
Percent Purity 99%
Chemical Name or Material Chlorpheniramine maleate
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-037-5
RTECSNumber US6503000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.