Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorpheniramine Maleate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601638 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
50 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-638 25 g Amber Glass Bottle
18-601-639 100 g Amber Glass Bottle
18-601-640 1 kg Amber Glass Bottle
18-601-641 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-638 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CH13325GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

CH133, 113-92-8, C16H19CIN2·C4H4O4

Spectrum™ Chemical Chlorpheniramine Maleate, USP is a first generation alkylamine antihistamine that is prevents the appearance of allergic conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 113-92-8
Molecular Formula C20H23ClN2O4
Molecular Weight (g/mol) 390.86
MDL Number MFCD00069225
InChI Key DBAKFASWICGISY-BTJKTKAUNA-N
IUPAC Name [3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylazanium (2Z)-3-carboxyprop-2-enoate
SMILES OC(=O)\C=C/C([O-])=O.C[NH+](C)CCC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation -0.10° to +0.10°
Percent Purity 98-102%
Grade USP
Physical Form Powder or Waxy Solid
Chemical Name or Material Chlorpheniramine Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.