Learn More
Medchemexpress LLC Chlorphenoxamine | 77-38-3 | 99.2% | 10 MM * 1 ML
SDP
Catalog No. 5000411095
Encompass_Preferred
Chlorphenoxamine is an antihistamine and anticholinergic agent that acts as a GPCR antagonist. It has been shown to inhibit several lethal viral diseases, including SARS-CoV, MERS-CoV, EBOV, and malaria. It exhibits anti-filovirus activity against EBOV and Marburg virus (MARV) with IC50s of 1.1 μM and 6.2 μM, respectively.
  • Antihistamine and anticholinergic agent.
  • GPCR antagonist.
  • Inhibits multiple lethal viral diseases (SARS-CoV, MERS-CoV, EBOV, malaria).
  • Anti-filovirus activity against EBOV (IC50 of 1.1 μM) and Marburg virus (MARV) (IC50 of 6.2 μM).
  • Used for allergic conditions, urticaria, viral diseases, and Parkinson's disease.
Catalog No. 50-004-11095 Supplier Medchemexpress LLC Supplier No. HYB160710MM/1ML
Chlorphenoxamine is an antihistamine and anticholinergic agent that acts as a GPCR antagonist. It has been shown to inhibit several lethal viral diseases, including SARS-CoV, MERS-CoV, EBOV, and malaria. It exhibits anti-filovirus activity against EBOV and Marburg virus (MARV) with IC50s of 1.1 μM and 6.2 μM, respectively.
  • Antihistamine and anticholinergic agent.
  • GPCR antagonist.
  • Inhibits multiple lethal viral diseases (SARS-CoV, MERS-CoV, EBOV, malaria).
  • Anti-filovirus activity against EBOV (IC50 of 1.1 μM) and Marburg virus (MARV) (IC50 of 6.2 μM).
  • Used for allergic conditions, urticaria, viral diseases, and Parkinson's disease.