Learn More
Cayman Chemical Chlorphenesin Carbamate, Purity: ≥98%, CAS: 886-74-8, 100 mg
SDP
Catalog No. 502919107
Encompass_Preferred
A muscle relaxant; decreases the dorsal root potential induced by a current-stimulated ventral root, as well as L-glutamate-induced motor neuron discharges, in isolated frog spinal cords at 1 mM; reduces the amplitude of polysynaptic responses in ipsilateral ventral root L5 induced by electrical stimulation of dorsal root L5 in spinalized rats at 50 mg/kg; decreases monosynaptic signal transfer from primary afferent fibers to motor neurons in the same animals at 50 mg/kg
Catalog No. 50-291-9107 Supplier Cayman Chemical Supplier No. 39252100 MG
A muscle relaxant; decreases the dorsal root potential induced by a current-stimulated ventral root, as well as L-glutamate-induced motor neuron discharges, in isolated frog spinal cords at 1 mM; reduces the amplitude of polysynaptic responses in ipsilateral ventral root L5 induced by electrical stimulation of dorsal root L5 in spinalized rats at 50 mg/kg; decreases monosynaptic signal transfer from primary afferent fibers to motor neurons in the same animals at 50 mg/kg