Learn More
Selleck Chemical LLC Chlorpromazine 25mg 50-53-3 CPZ
SDP
Supplier:  Selleck Chemical LLC S574925MG
Encompass_Preferred
Chlorpromazine (CPZ) is a low-potency typical antipsychotic agent that is used to treat psychotic disorders such as schizophrenia. Chlorpromazine (CPZ) inhibits 5-HT(2A) receptor and dopamine D2 in vitro. Chlorpromazine (CPZ) also inhibits sodium channel and blocks HERG potassium channels with IC50 of 21.6 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8207