Learn More
Medchemexpress LLC Chlorpromazine-d6 hydrochloride | 1228182-46-4 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0407AS1MG
Encompass_Preferred
Chlorpromazine-d6 (hydrochloride) is the deuterium labeled Chlorpromazine. Chlorpromazine is an inhibitor of dopamine receptor, 5-HT receptor, potassium channel, and sodium channel. It is used for research purposes only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Chlorpromazine
  • Inhibits dopamine receptor, 5-HT receptor, potassium channel, and sodium channel
  • Used as a tracer or internal standard for quantitative analysis
  • Deuteration can affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-07037