Learn More
Medchemexpress LLC Chlorpromazine-d6 (oxalate) | 1276197-23-9 | C19H15D6CIN2O4S | 1 MG

Supplier: Medchemexpress LLC HY151098S1MG
Chlorpromazine-d6 oxalate is a stable isotope of Chlorpromazine, presented as a white to off-white solid. This compound is primarily used as a laboratory chemical for research purposes and in the manufacture of various substances. It is characterized by its high purity and isotopic enrichment, ensuring reliability for sensitive experimental applications.
- Stable under recommended storage conditions
- For research use only
- Appearance: White to off-white solid
- Purity (HPLC): 99%
- Isotopic enrichment: 99.4%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.