Learn More
Apexbio Technology LLC Chlorpromazine HCl 69-09-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062357
Encompass_Preferred
Chlorpromazine HCl (CAS 69-09-0) is a phenothiazine-based dopamine receptor antagonist with significant activity at G protein-coupled dopamine receptors in the central nervous system Mechanistically chlorpromazine directly binds to dopamine receptors competitively inhibiting receptor-ligand interactions as evidenced by its inhibition of [ H]spiperone binding in vitro In vivo studies indicate chronic chlorpromazine administration in rats induces behavioral sensitization and altered motor responses linked to DA and NMDA receptor function Clinically chlorpromazine is established as a conventional antipsychotic approved for schizophrenia treatment making it valuable in neuropharmacology research
Catalog No. 50-000-62357 Supplier Apexbio Technology LLC Supplier No. B14805.1
Chlorpromazine HCl (CAS 69-09-0) is a phenothiazine-based dopamine receptor antagonist with significant activity at G protein-coupled dopamine receptors in the central nervous system Mechanistically chlorpromazine directly binds to dopamine receptors competitively inhibiting receptor-ligand interactions as evidenced by its inhibition of [ H]spiperone binding in vitro In vivo studies indicate chronic chlorpromazine administration in rats induces behavioral sensitization and altered motor responses linked to DA and NMDA receptor function Clinically chlorpromazine is established as a conventional antipsychotic approved for schizophrenia treatment making it valuable in neuropharmacology research