Learn More
Cayman Chemical Chlorpropamide, Purity: ≥98%, CAS: 94-20-2, 50 g
SDP
Catalog No. 502936267
Encompass_Preferred
A first generation sulfonylurea; increases glycolysis and inhibits glucagon-induced gluconeogenesis in isolated rat hepatocytes at 0.2 mM; reverses increases in blood glucose levels induced by hydrochlorothiazide and previous insulin in a rabbit model of diabetes at 250 mg/kg once or 10 mg/kg per day for six days
Catalog No. 50-293-6267 Supplier Cayman Chemical Supplier No. 2608750 G
A first generation sulfonylurea; increases glycolysis and inhibits glucagon-induced gluconeogenesis in isolated rat hepatocytes at 0.2 mM; reverses increases in blood glucose levels induced by hydrochlorothiazide and previous insulin in a rabbit model of diabetes at 250 mg/kg once or 10 mg/kg per day for six days