Learn More
Medchemexpress LLC Chlorpropamide | 94-20-2 | 25 MG
SDP
Catalog No. 5000414262
Encompass_Preferred
Chlorpropamide is an oral hypoglycemic agent that can influence acetaldehyde metabolism in mice and has a potentiating effect on antidiuretic hormone. It increases the cAMP accumulation in the thick ascending limb of the loop of Henle (MAL) in vitro.
  • Enhances the antidiuretic effect of vasopressin in rats.
  • Lowers blood sugar levels in rats.
  • Influences acetaldehyde metabolism in mice.
Catalog No. 50-004-14262 Supplier Medchemexpress LLC Supplier No. 25MGHYB1429R
Chlorpropamide is an oral hypoglycemic agent that can influence acetaldehyde metabolism in mice and has a potentiating effect on antidiuretic hormone. It increases the cAMP accumulation in the thick ascending limb of the loop of Henle (MAL) in vitro.
  • Enhances the antidiuretic effect of vasopressin in rats.
  • Lowers blood sugar levels in rats.
  • Influences acetaldehyde metabolism in mice.