Learn More
Medchemexpress LLC Chlorpropamide | 94-20-2 | 99.8% | 250 MG
SDP
Catalog No. 5000414487
Encompass_Preferred
Chlorpropamide is an oral hypoglycemic agent that can influence acetaldehyde metabolism in mice and has a potentiating effect on antidiuretic hormone. This product is for research use only and not sold to patients.
  • Oral hypoglycemic agent
  • Influences acetaldehyde metabolism in mice
  • Potentiates antidiuretic hormone
  • High purity (99.8%)
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO
  • Insoluble in water
Catalog No. 50-004-14487 Supplier Medchemexpress LLC Supplier No. 250MGHYB1429R
Chlorpropamide is an oral hypoglycemic agent that can influence acetaldehyde metabolism in mice and has a potentiating effect on antidiuretic hormone. This product is for research use only and not sold to patients.
  • Oral hypoglycemic agent
  • Influences acetaldehyde metabolism in mice
  • Potentiates antidiuretic hormone
  • High purity (99.8%)
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO
  • Insoluble in water