Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorpropamide (standard) | 94-20-2 | 98.7% | 100 MG
SDP

Catalog No. 5000412952
Encompass_Preferred

Chlorpropamide (Standard) is an analytical standard used in research and analytical applications. This product functions as an oral hypoglycemic agent and is studied in non-insulin-dependent diabetes mellitus (NIDDM).

  • Analytical standard for Chlorpropamide
  • Intended for research and analytical applications
  • Functions as an oral hypoglycemic agent
  • Studied in non-insulin-dependent diabetes mellitus (NIDDM)
  • Chemical formula: C10H13ClN2O3S
  • Molecular weight: 276.74
  • Research area: Endocrinology
  • Targets: Na+/K+ ATPase, Reference Standards

Catalog No. 50-004-12952 Supplier Medchemexpress LLC Supplier No. HYB1429R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorpropamide (Standard) is an analytical standard used in research and analytical applications. This product functions as an oral hypoglycemic agent and is studied in non-insulin-dependent diabetes mellitus (NIDDM).

  • Analytical standard for Chlorpropamide
  • Intended for research and analytical applications
  • Functions as an oral hypoglycemic agent
  • Studied in non-insulin-dependent diabetes mellitus (NIDDM)
  • Chemical formula: C10H13ClN2O3S
  • Molecular weight: 276.74
  • Research area: Endocrinology
  • Targets: Na+/K+ ATPase, Reference Standards

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.